Learn More
Diagnostic Biosystems PEROXIDASE Universal HRP Immunostaining Kit (For Mouse and Rabbit Primary Antibodies)
Catalog No. 502407564
Encompass_Preferred
This product is intended for qualitative immunohistochemistry with normal and neoplastic formalin-fixed, paraffin-embedded tissue sections, tobe viewed by light microscopy. Clinical interpretation of staining results should be accompanied by histological studies with proper controls. Patients’ clinical histories and other relevant diagnostic tests should be utilized by a qualified person (s) when evaluating and interpreting results. This kit is designed to label specific primary antibodies immunohistochemically on tissue sections. The immunohistochemical protocol defined in this brochure is just a guideline. Depending upon tissue fixation conditions, the primary antibody employed, and the user's experience, we encourage individual laboratory to optimize its own protocol. These reagents were tested and quality controlled using tissue sections, however, they can also be optimized for cell smears and cytospin preparations. Sufficient reagents are provided to run 50 to 100 tests.
Catalog No. 50-240-7564 Supplier Diagnostic Biosystems Supplier No. KP50
This product is intended for qualitative immunohistochemistry with normal and neoplastic formalin-fixed, paraffin-embedded tissue sections, tobe viewed by light microscopy. Clinical interpretation of staining results should be accompanied by histological studies with proper controls. Patients’ clinical histories and other relevant diagnostic tests should be utilized by a qualified person (s) when evaluating and interpreting results. This kit is designed to label specific primary antibodies immunohistochemically on tissue sections. The immunohistochemical protocol defined in this brochure is just a guideline. Depending upon tissue fixation conditions, the primary antibody employed, and the user's experience, we encourage individual laboratory to optimize its own protocol. These reagents were tested and quality controlled using tissue sections, however, they can also be optimized for cell smears and cytospin preparations. Sufficient reagents are provided to run 50 to 100 tests.