Learn More
Biocare Medical, LLC intelliPATH FLX Peroxidase Blocking Reagent - 20 ml
SDP
Catalog No. NC1416521
Encompass_Preferred
Peroxidazed Blocking Reagent is a highly stable form of hydrogen peroxide for blocking endogenous peroxidase. It is very effective for blocking non-specific staining in red blood cells. It is non-flammable, safer and less toxic when compared to hydrogen peroxide/methanol formulations.
This item is not returnable. View return policy
Catalog No. NC1416521 Supplier Biocare Medical, LLC Supplier No. IPB5000G20
Peroxidazed Blocking Reagent is a highly stable form of hydrogen peroxide for blocking endogenous peroxidase. It is very effective for blocking non-specific staining in red blood cells. It is non-flammable, safer and less toxic when compared to hydrogen peroxide/methanol formulations.