Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Goat Anti-Mouse IgG, F(ab')₂ fragment specific, 1.5ml
SDP
Catalog No. NC9131220
Encompass_Preferred
Peroxidase-AffiniPure Goat Anti-Mouse IgG, F(ab')2 Fragment Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9131220 Supplier Jackson Immuno Research Labs Supplier No. 115035072
Peroxidase-AffiniPure Goat Anti-Mouse IgG, F(ab')2 Fragment Specific (min X Hu,Bov,Hrs Sr Prot)