Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (GOAT) Antibody Peroxidase Conjugated (Min X Bv Ch Gt GP Ham Hs Hu Rb Rt & Sh Serum Proteins) - 610-103-121
SDP
Supplier:  Rockland Immunochemicals 610103121
Encompass
Peroxidase Secondary antibody reagents are ideal for western blotting, Immunohistochemistry and ELISA as well as other antibody detection methods.
Catalog No. NC9575145