Learn More
Crescent Chemical Co Inc PEROXIDASE FROM HORSERADISH
SDP
Catalog No. 50247995
Encompass_Preferred
Peroxidase from horseradish min. 1 000 U/mg lyophil. For the determination of peroxide (1). Used as an indicator enzyme in reactions where peroxide is produced (2). For labelling antibodies in ELISA (3, 4). RZ (= A 403/A 275) = 3.0. Size - 100MG Storage Conditions - +2 °C TO +8 °C
Catalog No. 50-247-995 Supplier Crescent Chemical Co Inc Supplier No. SE31941.03
Peroxidase from horseradish min. 1 000 U/mg lyophil. For the determination of peroxide (1). Used as an indicator enzyme in reactions where peroxide is produced (2). For labelling antibodies in ELISA (3, 4). RZ (= A 403/A 275) = 3.0. Size - 100MG Storage Conditions - +2 °C TO +8 °C