Learn More
Worthington Biochemical Corporation PEROXIDASE HORSERADISH 100MG
SDP
Catalog No. NC9156828
Encompass
A soluble, dialyzed, lyophilized powder. RZ > or = 1.0.
Catalog No. NC9156828 Supplier Worthington Biochemical Corporation Supplier No. LS002559
A soluble, dialyzed, lyophilized powder. RZ > or = 1.0.