Learn More
Aat Bioquest Amplite™ Colorimetric Peroxidase (HRP) Assay Kit *Blue Color*
SDP
Catalog No. 501953362
Encompass_Preferred
Peroxidase is a small molecule (MW ~40 KD) that can usually be conjugated to an antibody in a 4:1 ratio. Due to its small size, it rarely causes steric hindrance problem with antibody/antigen complex formation.
Catalog No. 50-195-3362 Supplier Aat Bioquest Supplier No. 11551
Peroxidase is a small molecule (MW ~40 KD) that can usually be conjugated to an antibody in a 4:1 ratio. Due to its small size, it rarely causes steric hindrance problem with antibody/antigen complex formation.