Learn More
Biocare Medical, LLC Peroxidazed 1 - 500 ml
SDP
Catalog No. 5083251
Encompass_Preferred
Peroxidazed 1 is a highly stable form of hydrogen peroxide for blocking endogenous peroxidase. It is very effective for blocking non-specific staining in red blood cells. It is non-flammable, safer and less toxic when compared to hydrogen peroxide/methanol formulations. It can also be used on an automated staining system.
Catalog No. 50-832-51 Supplier Biocare Medical, LLC Supplier No. PX968M
Peroxidazed 1 is a highly stable form of hydrogen peroxide for blocking endogenous peroxidase. It is very effective for blocking non-specific staining in red blood cells. It is non-flammable, safer and less toxic when compared to hydrogen peroxide/methanol formulations. It can also be used on an automated staining system.