Learn More
Biocare Medical, LLC PEROXIDAZED DILUENT 125ML
SDP
Catalog No. 5083253
Encompass_Preferred
Perozidazed Diluent may be used to dilute Peroxidazed 1, as some frozen tissues or delicate antigens require different concentrations of hydrogen peroxide for blocking endogenous peroxidase.
Catalog No. 50-832-53 Supplier Biocare Medical, LLC Supplier No. PX970LH
Perozidazed Diluent may be used to dilute Peroxidazed 1, as some frozen tissues or delicate antigens require different concentrations of hydrogen peroxide for blocking endogenous peroxidase.