Learn More
Biogenex Laboratories Peroxide Blocks
SDP
Catalog No. NC0508345
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Endogenous peroxidase is most commonly encountered in red blood cells, kidney and liver tissues. When Horseradish Peroxidase (HRP) is used as the labeling enzyme, this peroxide block should be used prior to the application of primary antibody.
Catalog No. NC0508345 Supplier Biogenex Laboratories Supplier No. HK1115K
This item has a minimum qty of 2 per supplier requirements.
Endogenous peroxidase is most commonly encountered in red blood cells, kidney and liver tissues. When Horseradish Peroxidase (HRP) is used as the labeling enzyme, this peroxide block should be used prior to the application of primary antibody.