Learn More
MSC PEROXIDE CLNR CONCENTRATE
Catalog No. 502818659
Encompass
The 3M™ Peroxide Cleaner Concentrate 34 is an all-purpose, light-duty peroxide cleaner concentrate. It is a versatile, daily-use cleaner effective against stains and soils. This low-odor product leaves no messy residue behind. It uses effervescing action to clean carpet and non-porous surfaces such as floors, walls, tables, chairs, countertops, bathroom fixtures, sinks, bed frames, doors, shelves, tile and grout, and more.
  • Automatic dispensing eliminates measuring and mixing and helps ensure accurate dilutions
  • No rinsing is required, saving time and labor
  • Designed to be used with cold water for enhanced user safety and energy savings
  • Green Seal GS-37 certification
  • Versatile, daily-use cleaner effective against stains and soils
  • Low odor product leaves no messy residue behind
  • Uses effervescing action to clean carpet and non-porous surfaces
Catalog No. 50-281-8659 Supplier MSC Supplier No. 11565108
The 3M™ Peroxide Cleaner Concentrate 34 is an all-purpose, light-duty peroxide cleaner concentrate. It is a versatile, daily-use cleaner effective against stains and soils. This low-odor product leaves no messy residue behind. It uses effervescing action to clean carpet and non-porous surfaces such as floors, walls, tables, chairs, countertops, bathroom fixtures, sinks, bed frames, doors, shelves, tile and grout, and more.
  • Automatic dispensing eliminates measuring and mixing and helps ensure accurate dilutions
  • No rinsing is required, saving time and labor
  • Designed to be used with cold water for enhanced user safety and energy savings
  • Green Seal GS-37 certification
  • Versatile, daily-use cleaner effective against stains and soils
  • Low odor product leaves no messy residue behind
  • Uses effervescing action to clean carpet and non-porous surfaces