Learn More
Cole-Parmer Water Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottle
Catalog No. NC0330796 Shop All Cole-Parmer Products
Encompass_Preferred
ITS 480014 Water Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottleObtain test results within 90 secondsTest strips feature bright easy-to-read colorsNo hazardous reagentsThese economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
Catalog No. NC0330796 Supplier Cole-Parmer Supplier No. 9953130
ITS 480014 Water Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottleObtain test results within 90 secondsTest strips feature bright easy-to-read colorsNo hazardous reagentsThese economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.