Learn More
Serim Research Corp Serim GUARDIAN™ Residual Peroxide Test Strips
SDP
Catalog No. NC1766186
Encompass_Preferred
Serim GUARDIAN™ Residual Peroxide Test Strips provide a rapid and convenient means for measuring the concentration of peroxide remaining in dialysate lines and dialyzers after disinfection with peracetic acid/peroxide-based disinfectants. Kit contains 6 bottles of 100 tests
Catalog No. NC1766186 Supplier Serim Research Corp Supplier No. 5105
Serim GUARDIAN™ Residual Peroxide Test Strips provide a rapid and convenient means for measuring the concentration of peroxide remaining in dialysate lines and dialyzers after disinfection with peracetic acid/peroxide-based disinfectants. Kit contains 6 bottles of 100 tests