Learn More
Sigma Aldrich Fine Chemicals Biosciences PeroxiDetectTM Kit
Catalog No. 502904648
Encompass
The PeroxiDetect Kit for the Determination of Aqueous and Lipid Hydroperoxides was developed for the measurement of peroxides in biological systems which is an important factor in determining the degree of free radicals present in specific tissues.
Catalog No. 50-290-4648 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PD11KT
The PeroxiDetect Kit for the Determination of Aqueous and Lipid Hydroperoxides was developed for the measurement of peroxides in biological systems which is an important factor in determining the degree of free radicals present in specific tissues.