Learn More
Sino Biological Peroxiredoxin 5 / PRDX5 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11263R125100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Peroxiredoxin 5 / PRDX5 (rh Peroxiredoxin 5 / PRDX5; Catalog#11263-H07E; P30044-2; Met53-Leu214).
Catalog No. 50-224-7871