Learn More
Cayman Chemical Peroxynitrite, CAS: 14042-01-4, 1 mL
SDP
Catalog No. 502894644
Encompass_Preferred
A highly ROS formed in vivo by the reaction of NO with superoxide; acts as a powerful oxidizing agent that can initiate lipid peroxidation, oxidize sulfhydryls, and nitrate the aromatic residues of proteins
Catalog No. 50-289-4644 Supplier Cayman Chemical Supplier No. 815651 ML
A highly ROS formed in vivo by the reaction of NO with superoxide; acts as a powerful oxidizing agent that can initiate lipid peroxidation, oxidize sulfhydryls, and nitrate the aromatic residues of proteins