Learn More
Sino Biological PERP1 / MZB1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 15682R022100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PERP1 / MZB1 (rh PERP1 / MZB1; Catalog#15682-H08H; Q8WU39-1; Met 1- Thr 185).
Catalog No. 50-224-7783