Learn More
Sino Biological PERP1 / MZB1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 15682T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PERP1 / MZB1 (rh PERP1 / MZB1; Catalog#15682-H08H; Q8WU39-1; Met 1- Thr 185). PERP1 / MZB1 specific IgG was purified by Human PERP1 / MZB1 affinity chromatography.
Catalog No. 50-224-7570