Learn More
Cayman Chemical Perphenazine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 155593-75-2, 5 mg
SDP
Catalog No. 502946490
Encompass_Preferred
An internal standard for the quantification of perphenazine by GC- or LC-MS
Catalog No. 50-294-6490 Supplier Cayman Chemical Supplier No. 332985 MG
An internal standard for the quantification of perphenazine by GC- or LC-MS