Learn More
Cayman Chemical PerphenazIn SulfoxIde 10mg
SDP
Catalog No. 502912424
Encompass_Preferred
An active metabolite of perphenazine; selectively binds to dopamine D2 and α1-ARs over α2-ARs (Kis= 5.9, 24, and 683 nM, respectively) in rat brain; increases thrombin-induced increases in phosphatidylinositol levels in isolated human platelets at 5 µM
Catalog No. 50-291-2424 Supplier Cayman Chemical Supplier No. 3580110 MG
An active metabolite of perphenazine; selectively binds to dopamine D2 and α1-ARs over α2-ARs (Kis= 5.9, 24, and 683 nM, respectively) in rat brain; increases thrombin-induced increases in phosphatidylinositol levels in isolated human platelets at 5 µM