Learn More
Apexbio Technology LLC Perphenazine 58-39-9 1g
SDP
Catalog No. 502240554
Encompass_Preferred
Perphenazine (CAS 58-39-9) is a phenothiazine derivative that primarily acts as an antagonist at dopamine D receptors with dissociation constant (Ki) of 1 4 nM and also displays significant binding to histamine H (Ki 8 nM) A-adrenergic (Ki 10 nM) A- B- C-adrenergic and muscarinic M (Ki 1848 nM) receptors Its antagonism at D receptors underlies its ability to modulate dopaminergic neurotransmission while interaction with additional receptor subtypes may account for its effects on the central nervous system and antiemetic properties Perphenazine is widely utilized in research investigating mechanisms of antipsychotic activity dopaminergic regulation and drug-induced cell death pathways
This item is not returnable. View return policy
Catalog No. 50-224-0554 Supplier Apexbio Technology LLC Supplier No. B61571000
Perphenazine (CAS 58-39-9) is a phenothiazine derivative that primarily acts as an antagonist at dopamine D receptors with dissociation constant (Ki) of 1 4 nM and also displays significant binding to histamine H (Ki 8 nM) A-adrenergic (Ki 10 nM) A- B- C-adrenergic and muscarinic M (Ki 1848 nM) receptors Its antagonism at D receptors underlies its ability to modulate dopaminergic neurotransmission while interaction with additional receptor subtypes may account for its effects on the central nervous system and antiemetic properties Perphenazine is widely utilized in research investigating mechanisms of antipsychotic activity dopaminergic regulation and drug-induced cell death pathways