Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Perphenazine-d4 (phenothiazine-1,3,7,9-d4) | 155593-75-2 | 99.0% | 10 MG
SDP

Catalog No. 5000298418
Encompass_Preferred

Perphenazine-d4 is the deuterium-labeled analog of perphenazine supplied as a research reference standard for use as an internal standard in LC-MS/MS and other quantitative mass spectrometry assays. It is provided as a high-purity powder for analytical and method development applications.

  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High purity (99.0%) for accurate quantitation.
  • Packaged as a 10 mg powder for analytical use.
  • Chemical formula C21H22D4ClN3OS; molecular weight 407.99 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-98418 Supplier Medchemexpress LLC Supplier No. HYA0077S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perphenazine-d4 is the deuterium-labeled analog of perphenazine supplied as a research reference standard for use as an internal standard in LC-MS/MS and other quantitative mass spectrometry assays. It is provided as a high-purity powder for analytical and method development applications.

  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High purity (99.0%) for accurate quantitation.
  • Packaged as a 10 mg powder for analytical use.
  • Chemical formula C21H22D4ClN3OS; molecular weight 407.99 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.