Learn More
Medchemexpress LLC Perphenazine sulfoxide | 10078-25-8 | MFCD01702800 | 95.0% | 419.97 g/mol | C21H26ClN3O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1330925MG
Encompass_Preferred
Perphenazine sulfoxide is the sulphoxide metabolite of the antipsychotic perphenazine, formed by oxidation. Supplied as an analytical standard for laboratory research and analytical applications, this compound is intended for research use only and not for human or veterinary use. Common applications include metabolite identification, method development, and use as a reference material. CAS 10078-25-8; molecular weight 419.97 g/mol; formula C21H26ClN3O2S.
  • Analytical standard suitable for chromatographic and spectrometric methods.
  • Useful for metabolite identification and method validation.
  • Provided as a solid reference material for laboratory use.
  • Accompanied by quality documentation such as a certificate of analysis when available.
  • Available in small weighed quantities appropriate for analytical workflows.
Catalog No. 50-002-62542