Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perrhenic Acid Solution

Catalog No. 2619635G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-196-35G 5 g
26-196-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 26-196-35G Supplier Sigma Aldrich Supplier No. 2619635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

It is also used to prepare highly conducting and superconducting synthetic metals.

Specifications

CAS 13768-11-1
Density 2.16 g/mL at 25°C
Molecular Formula HO4Re
Linear Formula HReO4
MDL Number MFCD00011326
Quantity 5 g
Synonym Hydrogen perrhenate; Hydrogen tetraoxorhenate
Molecular Weight (g/mol) 251.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.