Learn More
Medchemexpress LLC Persephin Human 10ug
SDP
Supplier:  Medchemexpress LLC HYP7103410UG
Encompass_Preferred
Recombinant human Persephin is a tag-free, E. coli-expressed neurotrophic protein supplied as a lyophilized powder. It forms a disulfide-linked homodimer that selectively supports midbrain dopaminergic and motor neuron survival and function. The product offers high purity and low endotoxin for research applications.
  • Tag-free, E. coli-expressed recombinant human protein
  • Disulfide-linked homodimer with neurotrophic activity for dopaminergic and motor neurons
  • High purity (>95%) as determined by reducing SDS-PAGE
  • Low endotoxin level (<1 EU/μg) measured by LAL
  • Lyophilized formulation from 4 mM HCl for stability
  • Stable when stored at -20°C; aliquot and freeze for long-term storage
  • Reconstitute at ≥100 μg/mL and add carrier protein for long-term storage
Catalog No. 50-001-85073