Learn More
Medtec Medical Inc PASS 2™ Personal Aseptic Sampling System™, 5 pk, Trypticase Soy Broth (TSB), 100 mL bag, 20 mL vials, 3 mL ampules, red dye
SDP
Catalog No. NC1186655
Encompass
The PASS 2 Kit Personal Aseptic Sampling System is a comprehensive training and validation aid for evaluating aseptic technique in sterile compounding environments. This system is vital for continuous quality improvement in IV pharmacies, allowing for thorough testing of both manual and automated processes while reducing the risk of contamination. Adhering to United States Pharmacopeia (USP) standards, it incorporates convenient prepackaged media and organized documentation for optimal usability.
  • Aseptic technique validation for training and quality assurance
  • Minimizes contamination risk during testing
  • Complies with USP standards
  • Prepackaged media components for ease of use
  • Includes integrated log sheets for organized documentation
  • Delivered in safe, lint-free thermal form trays
  • Allows testing for up to 5 individuals per kit
  • Supports continuous quality improvement in IV pharmacies
This item is not returnable. View return policy
Catalog No. NC1186655 Supplier Medtec Medical Inc Supplier No. TBVA123
The PASS 2 Kit Personal Aseptic Sampling System is a comprehensive training and validation aid for evaluating aseptic technique in sterile compounding environments. This system is vital for continuous quality improvement in IV pharmacies, allowing for thorough testing of both manual and automated processes while reducing the risk of contamination. Adhering to United States Pharmacopeia (USP) standards, it incorporates convenient prepackaged media and organized documentation for optimal usability.
  • Aseptic technique validation for training and quality assurance
  • Minimizes contamination risk during testing
  • Complies with USP standards
  • Prepackaged media components for ease of use
  • Includes integrated log sheets for organized documentation
  • Delivered in safe, lint-free thermal form trays
  • Allows testing for up to 5 individuals per kit
  • Supports continuous quality improvement in IV pharmacies