Learn More
Med Vet International Personal Eyewash Station, Measures 8.75 × 4.25 × 2.25 in, J0989, Each
Catalog No. 5000585170
Encompass_Preferred
Mount on wall to provide immediate relief. This station holds two 32 oz. dosing bottles for thorough decontamination of face and eyes. Comfortable molded eye socket opening allows full flow of solution to completely irrigate affected eye. Imprinted emergency use instructions are on each bottle and station wall. Bottles are shipped empty to fill with water or saline depending on local regulations. Securely mount station with top and bottom holes (hardware included).
Catalog No. 50-005-85170 Supplier Med Vet International Supplier No. JOR989
Mount on wall to provide immediate relief. This station holds two 32 oz. dosing bottles for thorough decontamination of face and eyes. Comfortable molded eye socket opening allows full flow of solution to completely irrigate affected eye. Imprinted emergency use instructions are on each bottle and station wall. Bottles are shipped empty to fill with water or saline depending on local regulations. Securely mount station with top and bottom holes (hardware included).