Learn More
Brady™ Personal Protection (PPE) Labels - Self-Sticking Polyester, English and Spanish Bilingual
Catalog No. 19111799 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
EYE PROTECTION
Catalog No. Display Text
19-111-799 EYE PROTECTION
1 options
Catalog No. 19-111-799 Supplier Brady™ Supplier No. 49895
  • $31.75
    $22.65 / Pack of 5
    Save $9.10
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Personal Protection (PPE) Labels - Self-Sticking Polyester, English and Spanish Bilingual: Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Display Text EYE PROTECTION
Languages English, Spanish
Quantity 5/Pk.
Width (Metric) 92.71 mm