Learn More
GENEPATH DIAGNOSTICS INC Personalized genomics (PGx) from cheek swabs for isolation of human genomic DNA (RUO)
Catalog No. NC3949362
Encompass_Preferred
Supports preservation and stabilization of genomic nucleic acids along with direct addition into compatible PCR mixes (without the need for additional sample preparation or nucleic acid extraction). (RUO)
Catalog No. NC3949362 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10510410096
Supports preservation and stabilization of genomic nucleic acids along with direct addition into compatible PCR mixes (without the need for additional sample preparation or nucleic acid extraction). (RUO)