Learn More
AccuTec Blades™ Personna Plus™ Safety Scalpel System Disposable Scalpels
Catalog No. 22080073
Change view
Click to view available options
Size:
# 20
Catalog No. Size
22-080-073 # 20
1 options
Catalog No. 22-080-073 Supplier AccuTec Blades™ Supplier No. 739220
Scalpel features Personna Plus blade with MicroCoat™ edge for smooth, clean precision
  • Meets OSHA requirements
  • User never touches the blade
  • Full range of sizes available
  • New, single-use, transparent; with safety tab and thumb-notch
  • Radio-Opaque: x-ray detectable
  • Fully disposable
  • Fixed position for stability
  • Color coded by size
  • Sure Grip sides for easy opening, and closing
Compliance
Meets OSHA requirements
Specifications
Size # 20