Learn More
CHEMetrics™ by AquaPhoenix Persulfate Water Quality Visual Test Kit
SDP
Catalog No. 13299689
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-689 30
1 options
Catalog No. 13-299-689 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K7870
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Persulfate visual test kit supplied in a plastic case. Contains everything needed to perform 30 tests.
  • Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low and High Range Comparators, 25 mL sample cup, and instructions.
Specifications
Analytes Persulfate
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 5.6 and 7 to 70 ppm
Type Water Quality Visual Test Kit