Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pertussis Toxin

Catalog No. PHZ1174
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHZ1174 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHZ1174 Supplier Invitrogen™ Supplier No. PHZ1174
Add to Cart
Edge
Add to Cart

Formulation: Protein expressed in E. coli. Purity: >95%. Format: Lyophilized.

Islet-activating protein Pertussis toxin consists of an A protomer subunit (S1) which possesses both NAD+ glycohydrolase and ADP-ribosyltransferase activities, and B oligomer subunits (S2, S3, S4, and S5) which are responsible for attachment of the native toxin to eukaryotic cell surfaces. Pertussis toxin uncouples G proteins from receptors by ADP ribosylating a cysteine residue near the carboxyl terminus of the α subunit.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2665068
Purification Method PAGE
Quantity 50 μg
Content And Storage Store in refrigerator (2–8°C).
Inhibitor Properties Protein
Inhibitors GTPase Inhibitor
Shipping Condition Wet Ice
Specific Inhibit or Target G Proteins
Form Lyophilized
Purity or Quality Grade >95%
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.