Learn More
Enzo Life Sciences Pertussis toxin (Bordetella pertussis) (50 µg) CAS: 70323-44-3
SDP
Catalog No. NC9282261
Encompass_Preferred
Ultra-pure pertussis toxin delivering optimal antigenicity. Alternative name: Islet activating protein. PTX. Holotoxin. Purity: ≥95%. Formulation: Lyophilized. Containing sodium phosphate and sodium chloride. Source: From Bordetella pertussis, strain 165. Use/Stability: This preparation is lyophilized, aseptically packaged and sealed under vacuum. Do not vortex. Store at 4°C prior to and following reconstitution. Do not freeze. This toxin is considered to be biologically active. The lyophilized preparation will remain active for at least one year
Catalog No. NC9282261 Supplier Enzo Life Sciences Supplier No. BMLG1000050
 Restricted Item
Add to Cart
Add to Cart
Ultra-pure pertussis toxin delivering optimal antigenicity. Alternative name: Islet activating protein. PTX. Holotoxin. Purity: ≥95%. Formulation: Lyophilized. Containing sodium phosphate and sodium chloride. Source: From Bordetella pertussis, strain 165. Use/Stability: This preparation is lyophilized, aseptically packaged and sealed under vacuum. Do not vortex. Store at 4°C prior to and following reconstitution. Do not freeze. This toxin is considered to be biologically active. The lyophilized preparation will remain active for at least one year