Learn More
MilliporeSigma Pertussis Toxin, Bordetella pertussis, 50ug
Catalog No. 502306671
Encompass
Pertussis Toxin, Bordetella pertussis, catalyzes ADP-ribosylation of guanine nucleotide-binding regulatory proteins Gi, Go, and Gt. 70323-44-3, 50ug
Catalog No. 50-230-6671 Supplier MilliporeSigma Supplier No. 51656050UG
Pertussis Toxin, Bordetella pertussis, catalyzes ADP-ribosylation of guanine nucleotide-binding regulatory proteins Gi, Go, and Gt. 70323-44-3, 50ug