Learn More
Eurofins Discoverx PathHunter Pertuzumab Bioassay Kit, 2-Plate. (93-1042Y3-00095)
Catalog No. 502001911
Encompass
The PathHunter Pertuzumab Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses heregulin-β2-induced dimerization of ErbB2 and EGFR receptors. This product uses EFC technology. The cell line included overexpresses ErbB2 target protein fused to PK alongside ErbB3 target protein fused to EA. Upon ligand-induced dimerization of ErbB2 and ErbB3, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1911 Supplier Eurofins Discoverx Supplier No. 931042Y300095
The PathHunter Pertuzumab Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses heregulin-β2-induced dimerization of ErbB2 and EGFR receptors. This product uses EFC technology. The cell line included overexpresses ErbB2 target protein fused to PK alongside ErbB3 target protein fused to EA. Upon ligand-induced dimerization of ErbB2 and ErbB3, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.