Learn More
Genscript Corporation Antibody Kit Other Products Immunodetection 1/EA
Catalog No. 502529970
Encompass
GenScript Pertuzumab Immunogenicity Kit (Bridging ELISA) is designed for detection of anti-Pertuzumab antibody in serum and plasma samples. This kit utilizes a validated bridging immunoassay method based on the FDA EMA and NMPA Immunogenicity Guidelines
Catalog No. 50-252-9970 Supplier Genscript Corporation Supplier No. L00979
GenScript Pertuzumab Immunogenicity Kit (Bridging ELISA) is designed for detection of anti-Pertuzumab antibody in serum and plasma samples. This kit utilizes a validated bridging immunoassay method based on the FDA EMA and NMPA Immunogenicity Guidelines