Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the estimation of Pertuzumab (PERJETA) in serum and plasma. Assay Range: 40 – 2560 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBI1086
Encompass_Preferred
Pack Size: 1 x 96 wells. Introduction: Pertuzumab, sold under brand name PERJETA, is a monoclonal antibody used in combination with Trastuzumab and Docetaxel for the treatment of metastatic HER2-positive breast cancer. It is also used in the same combination as a neoadjuvant in early HER2-positive breast cancer.A first-in-class of a kind of drug called a HERdimerization inhibitor, Pertuzumab inhibits thedimerizationof HER2 with other HER receptors, which prevents them from signallingin ways that promote cell growth and proliferation. This mAb-based Pertuzumab ELISA accurately quantifies pertuzumab in human samples. If you are looking to use animal samples, please contact us. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <15%.
Catalog No. NC2747130