Learn More
Genscript Corporation Antibody Kit Other Products Immunodetection 1/EA
Catalog No. 502529971
Encompass
The Pertuzumab ELISA kit is a validated tool for whole Pertuzumab and its biosimilar quantification in biological matrices for drug research and development. GenScripts Pertuzumab Pharmacokinetic ELISA Kit had been comprehensively developed and validated for quantitative measurement of Pertuzumab in cynomolgus monkey serum and plasma based on the ICH M10 and the FDA bioanalytical method validation guidance for industry
Catalog No. 50-252-9971 Supplier Genscript Corporation Supplier No. L00978
The Pertuzumab ELISA kit is a validated tool for whole Pertuzumab and its biosimilar quantification in biological matrices for drug research and development. GenScripts Pertuzumab Pharmacokinetic ELISA Kit had been comprehensively developed and validated for quantitative measurement of Pertuzumab in cynomolgus monkey serum and plasma based on the ICH M10 and the FDA bioanalytical method validation guidance for industry