Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perylene Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022524 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2524 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2524 Supplier MilliporeSigma Supelco Supplier No. 45798100MG

missing translation for 'validMainframeMsg'

Perylene belongs to the class of polycyclic aromatic hydrocarbons. It is a hydrophobic, fluorescent probe used for imaging lipid bilayer membranes.

TRUSTED_SUSTAINABILITY

Specifications

CAS 198-55-0
Melting Point 276°C to 279°C (literature)
Molecular Formula C20H12
Linear Formula C20H12
MDL Number MFCD00004142
Quantity 100 mg
UN Number NONH for all modes of transport
Synonym Dibenz[de,kl]anthracene
Formula Weight 252.31
Grade Analytical Standard
Physical Form Neat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.