Learn More
Medchemexpress LLC Perylene-d12 | 1520-96-3 | 99.3% | 264.39 g·mol⁻¹ | C20D12 | 25 MG
SDP
Catalog No. 5000208768
Encompass_Preferred
Perylene-d12 is the deuterium-labeled isotopologue of perylene (C20D12), supplied as a solid for use as a stable isotope standard and tracer in analytical and research applications. It is high-purity and characterized by molecular weight 264.39 g·mol⁻¹ and CAS number 1520-96-3.
  • Deuterium-labeled perylene (C20D12).
  • High purity suitable for analytical standards.
  • Available in small milligram quantities for laboratory use.
  • Solid powder form with defined storage stability.
  • Documentation available: data sheet, COA, SDS.
Catalog No. 50-002-08768 Supplier Medchemexpress LLC Supplier No. 25MGHYW090294S
Perylene-d12 is the deuterium-labeled isotopologue of perylene (C20D12), supplied as a solid for use as a stable isotope standard and tracer in analytical and research applications. It is high-purity and characterized by molecular weight 264.39 g·mol⁻¹ and CAS number 1520-96-3.
  • Deuterium-labeled perylene (C20D12).
  • High purity suitable for analytical standards.
  • Available in small milligram quantities for laboratory use.
  • Solid powder form with defined storage stability.
  • Documentation available: data sheet, COA, SDS.