Learn More
Apexbio Technology LLC PERYLENE-D12-25MG
SDP
Catalog No. 5000570418
Encompass_Preferred
Perylene-d12 (CAS No 1520-96-3) is a deuterated chemical compound used primarily in the field of biomedical research as a tracer in mass spectrometry and as a standard for quantitative analysis Derived from perylene its mechanism of action involves intercalation into DNA and interaction with cellular membranes impacting nucleic acid processes Perylene-d12 exhibits potent in vitro activity typically observed in the nanomolar to low micromolar range It is widely used across various cell models and animal studies particularly in drug screening and toxicology assessments Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70418 Supplier Apexbio Technology LLC Supplier No. SL101825
Perylene-d12 (CAS No 1520-96-3) is a deuterated chemical compound used primarily in the field of biomedical research as a tracer in mass spectrometry and as a standard for quantitative analysis Derived from perylene its mechanism of action involves intercalation into DNA and interaction with cellular membranes impacting nucleic acid processes Perylene-d12 exhibits potent in vitro activity typically observed in the nanomolar to low micromolar range It is widely used across various cell models and animal studies particularly in drug screening and toxicology assessments Experimental concentrations typically depend on the specific research design