Learn More
Medchemexpress LLC HY-19168 5mg Medchemexpress, Perzinfotel CAS:144912-63-0 Purity:>98%
SDP
Catalog No. 501932828
Encompass_Preferred
Medchemexpress, HY-19168 5mg Perzinfotel CAS:144912-63-0 Perzinfotel (EAA-090) is a potent, selective, and competitive NMDA receptor antagonist with neuroprotective effects. Perzinfotel (EAA-090) shows high affinity ( IC 50 =30 nM) for the glutamate site [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2828 Supplier Medchemexpress LLC Supplier No. HY191685MG
Medchemexpress, HY-19168 5mg Perzinfotel CAS:144912-63-0 Perzinfotel (EAA-090) is a potent, selective, and competitive NMDA receptor antagonist with neuroprotective effects. Perzinfotel (EAA-090) shows high affinity ( IC 50 =30 nM) for the glutamate site [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.