Learn More
CELLTREAT Scientific Products PES Syringe Filter, 0.45m, 13mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229748
Encompass_Preferred
CELLTREAT Scientific Products 0.45¿m PES Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. PES (Polyethersulfone)-Most Popular Membrane. Low affinity for proteins and extractables. Faster flow rate than PVDF. Pre-filtration and filtration of buffers and culture media. Membrane size 13mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in peel-to-open paper/plastic blister packs. 75/case.
Catalog No. 50-202-063