Learn More
Sterlitech Corporation Polyethersulfone (PES) Membrane Filters, 0.2 Micron, 47mm, 100/Pk
SDP
Catalog No. NC1288573
Encompass_Preferred
Sterlitech PES (polyethersulfone) membrane filters, 0.2 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, low protein/drug binding, minimal extractables, and broad chemical tolerance; ideal for pharmaceutical, medical, and life science applications.
Catalog No. NC1288573 Supplier Sterlitech Corporation Supplier No. PES0247100
Sterlitech PES (polyethersulfone) membrane filters, 0.2 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, low protein/drug binding, minimal extractables, and broad chemical tolerance; ideal for pharmaceutical, medical, and life science applications.