Learn More
Sterlitech Corporation PES Syringe Filters, Sterlitech, 0.22 Micron, 25mm, 100/Pk
SDP
Supplier:  Sterlitech Corporation 1470357
Encompass_Preferred
PES Syringe Filter Applications:* Sterilize biological fluids, serum or tissue culture media additives* Sample preparation of aqueous solutions* High throughput, low binding filter* Units for sterile aqueous filtrations* Filter probe and hybridization solutions to reduce backgrounds* Sample preparation of protein-based* HPLC solutions* High throughputs when sterilizing or clarifying even the most viscous proteinaceous solutions
Catalog No. 50-195-2318