Learn More
Aviva Systems Biology Corp PES1 Antibody - C-terminal region (ARP55008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PES1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PES1. Alternate names: PESApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61394