Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 1,2,4-Trichlorobenzene, MilliporeSigma™ Supelco™

Catalog No. 111020618 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0618 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0618 Supplier MilliporeSigma Supelco Supplier No. 366271G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2,4-Trichlorobenzene can be used as a solvent for industrial purposes. It can also find applications as a termiticide and lubricant for household uses.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Ignition Point 1060°F
CAS 120-82-1
Vapor Pressure 1 mmHg (40°C)
Vapor Density >6 (vs. air)
Density 1.454 g/mL (at 25°C)
Flash Point 110°C
Physical Form Neat
Linear Formula C6H3 Cl3
MDL Number MFCD00000547
Melting Point 16°C
Boiling Point 214°C
Type Analytical Standard
Quantity 1 g
Formula Weight 181.45
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.