Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 1-Naphthaleneacetic acid, 95-100% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015713 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5713 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5713 Supplier MilliporeSigma Supelco Supplier No. 357451G
Add to Cart
Add to Cart

1-Naphthaleneacetic acid is widely employed in agriculture as a synthetic plant hormone in the auxin family or a plant growth regulator, which can find applications in tissue culture.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 86-87-3
Physical Form Neat
Linear Formula C12H10O2
MDL Number MFCD00004046
Synonym alpha-Naphthaleneacetic acid; 1-Naphthylacetic acid; NAA
Type Analytical Standard
Quantity 1 g
Formula Weight 186.21
Percent Purity 95 to 100% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.