Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 2,3-Dichloroaniline Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022470 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2470 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2470 Supplier MilliporeSigma Supelco Supplier No. 36701100MG

missing translation for 'validMainframeMsg'

Specifications

CAS 608-27-5
Melting Point 20°C to 25°C (literature)
Density 1.37 g/mL (at 20°C (literature))
Boiling Point 252°C (literature)
Flash Point 115°C (Closed Cup)
Molecular Formula C6H5Cl2N
Linear Formula Cl2C6H3NH2
Refractive Index n20/D 1.597 (literature)
Quantity 100 mg
MDL Number MFCD00007657
UN Number UN 3442 6.1 / PGII
Formula Weight 162.02
Grade Analytical
Physical Form Neat
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.