Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 2,4,5-Trichlorophenol, MilliporeSigma™ Supelco™

Catalog No. 111020332 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0332 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0332 Supplier MilliporeSigma Supelco Supplier No. 36513250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,5-Trichlorophenol is a phenolic organochloride, which is persistent as an environmental contaminant.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 95-95-4
Flash Point 133°C
Physical Form Neat
Linear Formula Cl3C6H2 OH
MDL Number MFCD00002170
Melting Point 67°C to 69°C
Boiling Point 248°C (740 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 197.45
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.